missing translation for 'onlineSavingsMsg'
Learn More

rac N,N-Didesmethyl-O-desmethyl Venlafaxine Glucuronide, TRC

Product Code. 30382926
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382926 1 mg 1mg
30382162 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382926 Supplier Toronto Research Chemicals Supplier No. TRCD4409751MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac N,N-Didesmethyl-O-desmethyl Venlafaxine Glucuronide
CAS 1799830-07-1
Molecular Formula C20H29NO8
Quantity 1 mg
InChI Formula InChI=1S/C20H29NO8/c21-10-13(20(27)8-2-1-3-9-20)11-4-6-12(7-5-11)28-19-16(24)14(22)15(23)17(29-19)18(25)26/h4-7,13-17,19,22-24,27H,1-3,8-10,21H2,(H,25,26)/t13?,14-,15-,16+,17-,19+/m0/s1
SMILES OC1(CCCCC1)C(CN)C2=CC=C(O[C@@H]([C@@H]3O)O[C@H](C(O)=O)[C@@H](O)[C@@H]3O)C=C2
Molecular Weight (g/mol) 411.45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.