missing translation for 'onlineSavingsMsg'
Learn More

rac N-Methyl Atomoxetine Oxalate, TRC

Product Code. 17962035
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962035 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962035 Supplier Toronto Research Chemicals Supplier No. TRCM288283100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 107688-86-8
Molecular Formula C18 H23 N O . C2 H2 O4
Quantity 100 mg
Synonym N,N-Ddimethyl-γ-(2-methylphenoxy)-benzenepropanamine Ethanedioate (1:1), rac N-Methyl Atomoxetine Oxalate
InChI Formula InChI=1S/C18H23NO.C2H2O4/c1-15-9-7-8-12-17(15)20-18(13-14-19(2)3)16-10-5-4-6-11-16;3-1(4)2(5)6/h4-12,18H,13-14H2,1-3H3;(H,3,4)(H,5,6)
SMILES CN(C)CCC(Oc1ccccc1C)c2ccccc2.OC(=O)C(=O)O
IUPAC Name N,N-dimethyl-3-(2-methylphenoxy)-3-phenyl-propan-1-amine;oxalic acid
Molecular Weight (g/mol) 359.42
Formula Weight 359.1733
Chemical Name or Material rac N-Methyl Atomoxetine Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.