missing translation for 'onlineSavingsMsg'
Learn More

rac N-Desethyl N-Benzyl Milnacipran Chloride, TRC

Product Code. 30357448
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357448 50 mg 50mg
30355230 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357448 Supplier Toronto Research Chemicals Supplier No. TRCD28905050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac N-Desethyl N-Benzyl Milnacipran Chloride
Analyte or Component Names rac N-Desethyl N-Benzyl Milnacipran Chloride
Shipping Condition Room Temperature
Molecular Formula C20 H25 N2 O . Cl
Synonym (1R,2S)-rel-2-(Aminomethyl)-N-ethyl-1-phenyl-N-(phenylmethyl)-cyclopropanecarboxamide Chloride
InChI Formula InChI=1S/C20H24N2O.ClH/c1-2-22(15-16-9-5-3-6-10-16)19(23)20(13-18(20)14-21)17-11-7-4-8-12-17;/h3-12,18H,2,13-15,21H2,1H3;1H/t18-,20+;/m1./s1
Solution Type Neat
SMILES [Cl-].CCN(Cc1ccccc1)C(=O)[C@@]2(C[C@@H]2C[NH3+])c3ccccc3
IUPAC Name [(1S,2R)-2-[benzyl(ethyl)carbamoyl]-2-phenylcyclopropyl]methylazanium;chloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 344.878
Quantity 50 mg
Formula Weight 344.166
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.