missing translation for 'onlineSavingsMsg'
Learn More

rac Metoprolol, TRC

Product Code. 30137660
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137660 1 g 1g
17939984 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137660 Supplier Toronto Research Chemicals Supplier No. TRCM3388151G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51384-51-1
Molecular Formula C15 H25 N O3

Specifications

Quantity 1 g
Synonym (RS)-Metoprolol, (±)-Metoprolol, 1-Isopropylamino-3-[4-(2-methoxyethyl)phenoxy]-2-propanol, 1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol, Beatrolol, CGP 2175, Meijoprolol, Metocard, Metohexal, Metoprolol, Seroken, Spesicor, Toprol, dl
InChI Formula InChI=1S/C15H25NO3/c1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3/h4-7,12,14,16-17H,8-11H2,1-3H3
SMILES COCCc1ccc(OCC(O)CNC(C)C)cc1
IUPAC Name 1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molecular Weight (g/mol) 267.364
Formula Weight 267.183
Percent Purity >95%
Chemical Name or Material Metoprolol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.