missing translation for 'onlineSavingsMsg'
Learn More

rac Metoprolol Hemi (+)-Tartrate, TRC

Product Code. 30041336
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041336 100 mg 100mg
18083940 50 mg 50mg
18062150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041336 Supplier Toronto Research Chemicals Supplier No. TRCM338790100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56392-17-7
Molecular Formula 2 C15 H25 N O3 . C4 H6 O6
Molecular Weight (g/mol) 684.81
Synonym Metoprolol Tartrate, 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, (2R,3R)-2,3-dihydroxybutanedioate (2:1), Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, (±)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1) (salt), 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, (2R,3R)-2,3-dihydroxybutanedioate (2:1) (salt) (9CI), (±)-1-(Isopropylamino)-3-[4-(2-methoxyethyl)phenoxy]-2-propanol tartrate, (±)-Metoprolol bitartrate, (±)-Metoprolol tartrate, 1-(Isopropylamino)-3-[4-(2-methoxyethyl)phenoxy]-2-propanol tartrate, 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1) (salt), Beloc, Betaloc, CGP 2175E, H 93/26, HCTCGP 2175E, Lopresor, Metapro, Metoprolol (+)-tartrate, Metoprolol tartrate, Metoprolol tartrate (2:1), Prelis, Seloken, Selopral, Slow Lopresor, Vasocardin
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
SMILES COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C15H25NO3.C4H6O6/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-1(3(7)8)2(6)4(9)10/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1
Formula Weight 684.3833
Chemical Name or Material Metoprolol Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.