missing translation for 'onlineSavingsMsg'
Learn More

rac Methotrimeprazine, TRC

Product Code. 30135230
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135230 25 mg 25mg
30012256 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135230 Supplier Toronto Research Chemicals Supplier No. TRCM26077525MG
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851-68-3
Molecular Formula C19H24N2OS
Molecular Weight (g/mol) 328.47
Synonym 10-[3-(Dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine, (±)-2-Methoxy-10-[3-(dimethylamino)-2-methylpropyl]phenothiazine, (±)-Methotrimeprazine, 10-[3-(Dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine, dl-Methotrimeprazine
IUPAC Name (2R)-3-(2-methoxyphenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
SMILES CN(C)CC(C)CN1C2=C(C=CC(OC)=C2)SC3=CC=CC=C31

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H24N2OS/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21/h5-11,14H,12-13H2,1-4H3/t14-/m1/s1
Formula Weight 328.1609
Chemical Name or Material rac Methotrimeprazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.