missing translation for 'onlineSavingsMsg'
Learn More

rac Ketorolac Tromethamine Salt, TRC

Product Code. 30135358
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135358 1 g 1g
30134283 5 g 5g
30005770 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135358 Supplier Toronto Research Chemicals Supplier No. TRCK2356501G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74103-07-4
Molecular Formula C15 H13 N O3 . C4 H11 N O3
Molecular Weight (g/mol) 376.4
Synonym Ketorolac Trometamol, 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, compd. with 2-amino-2-(hydroxymethyl)-1,3-propanediol (1:1), 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, (±)-, compd. with 2-amino-2-(hydroxymethyl)-1,3-propanediol (1:1), 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-, (±)-5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate (1:1) (salt), 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-, benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate (1:1) (salt) (9CI), Acular, Acular PF, Acuvail, Dolac, ISV 304, Ketanov, Ketolac, Ketorol, Ketorolac tromethamine, Lixidol, Tarazyn, Toradol, Toratex, Trometamol Ketorolac, ketorolac tris salt
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
SMILES NC(CO)(CO)CO.OC(=O)C1CCn2c1ccc2C(=O)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H13NO3.C4H11NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13;5-4(1-6,2-7)3-8/h1-7,11H,8-9H2,(H,18,19);6-8H,1-3,5H2
Formula Weight 376.1634
Percent Purity >95%
Chemical Name or Material Ketorolac trometamol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.