missing translation for 'onlineSavingsMsg'
Learn More

rac Ketorolac Ethyl Ester 6-Benzoyl Isomer, TRC

Product Code. 30383537
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383537 10 mg 10mg
30375493 1 mg 1mg
30386598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383537 Supplier Toronto Research Chemicals Supplier No. TRCE90008810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Ketorolac Ethyl Ester 6-Benzoyl Isomer
Analyte or Component Names rac Ketorolac Ethyl Ester 6-Benzoyl Isomer
Shipping Condition Room Temperature
Molecular Formula C17 H17 N O3
Synonym Ethyl 6-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate
InChI Formula InChI=1S/C17H17NO3/c1-2-21-17(20)14-8-9-18-11-13(10-15(14)18)16(19)12-6-4-3-5-7-12/h3-7,10-11,14H,2,8-9H2,1H3
Solution Type Neat
SMILES CCOC(=O)C1CCn2cc(cc12)C(=O)c3ccccc3
IUPAC Name ethyl 6-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 283.322
Quantity 10 mg
Formula Weight 283.121
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.