missing translation for 'onlineSavingsMsg'
Learn More

rac-Ketoprofen Tris Base Amide, TRC

Product Code. 30396459
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396459 100 mg 100mg
30394147 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396459 Supplier Toronto Research Chemicals Supplier No. TRCK200835100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Ketoprofen Tris Base Amide
CAS 1777830-05-3
Molecular Formula C20 H23 N O5
Quantity 100 mg
Synonym 3-Benzoyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]-α-methyl-benzeneacetamide
InChI Formula InChI=1S/C20H23NO5/c1-14(19(26)21-20(11-22,12-23)13-24)16-8-5-9-17(10-16)18(25)15-6-3-2-4-7-15/h2-10,14,22-24H,11-13H2,1H3,(H,21,26)
SMILES CC(C(=O)NC(CO)(CO)CO)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name 2-(3-benzoylphenyl)-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]propanamide
Molecular Weight (g/mol) 357.15762285
Formula Weight 357.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.