missing translation for 'onlineSavingsMsg'
Learn More

rac Ketoprofen Beta-D-Glucuronide Allyl Ester, TRC

Product Code. 30395891
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395891 1 mg 1mg
30396997 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395891 Supplier Toronto Research Chemicals Supplier No. TRCK2008301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Ketoprofen beta-D-Glucuronide Allyl Ester
Analyte or Component Names rac Ketoprofen beta-D-Glucuronide Allyl Ester
Shipping Condition Room Temperature
Molecular Formula C25H26O9
Synonym 1-(3-Benzoly-α-methylbenzeneacetate)-β-D-glucopyranuronic Acid Allyl Ester, dl-Ketoprofen Glucuronide Allyl Ester, Ketoprofen Allyl Glucuronate
InChI Formula InChI=1S/C25H26O9/c1-3-12-32-24(31)22-20(28)19(27)21(29)25(33-22)34-23(30)14(2)16-10-7-11-17(13-16)18(26)15-8-5-4-6-9-15/h3-11,13-14,19-22,25,27-29H,1,12H2,2H3/t14?,19?,20-,21?,22?,25-/m0/s1
Solution Type Neat
SMILES O[C@H](C(C(OCC=C)=O)O[C@@H](OC(C(C1=CC=CC(C(C2=CC=CC=C2)=O)=C1)C)=O)C3O)C3O
IUPAC Name allyl (3S,6S)-6-((2-(3-benzoylphenyl)propanoyl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 470.47
Quantity 1 mg
Formula Weight 470.16
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.