missing translation for 'onlineSavingsMsg'
Learn More

rac Ketoprofen-13C,d3, TRC

Product Code. 30399399
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399399 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399399 Supplier Toronto Research Chemicals Supplier No. TRCK2008021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Ketoprofen 13C,D3 (methyl 13C,d3)
CAS 1189508-77-7
Molecular Formula 13C C15 2H3 H11 O3
Quantity 1 mg
Synonym rac Ketoprofen-13C,d3, (±)-Ketoprofen 13C,D3 (methyl 13C,D3)
InChI Formula InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/i1+1D3
SMILES [2H][13C]([2H])([2H])C(C(=O)O)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name 2-(3-benzoylphenyl)-3,3,3-trideuterio(3^{13}C)propanoic acid
Molecular Weight (g/mol) 258.29
Formula Weight 258.1165
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.