missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac Indapamide-N-sulfonamido-Beta-D-glucuronide (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | rac Indapamide-N-sulfonamido-beta-D-glucuronide |
| Analyte or Component Names | rac Indapamide-N-sulfonamido-beta-D-glucuronide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H24 Cl N3 O9 S |
| Synonym | rac Indapamide-N-sulfonamido-β-D-glucuronide (Mixture of Diastereomers) |
| InChI Formula | InChI=1S/C22H24ClN3O9S/c1-10-8-11-4-2-3-5-14(11)26(10)24-20(30)12-6-7-13(23)15(9-12)36(33,34)25-21-18(29)16(27)17(28)19(35-21)22(31)32/h2-7,9-10,16-19,21,25,27-29H,8H2,1H3,(H,24,30)(H,31,32)/t10?,16-,17-,18+,19-,21+/m0/s1 |
| Solution Type | Neat |
| SMILES | CC1Cc2ccccc2N1NC(=O)c3ccc(Cl)c(c3)S(=O)(=O)N[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6R)-6-[[2-chloro-5-[(2-methyl-2,3-dihydroindol-1-yl)carbamoyl]phenyl]sulfonylamino]-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Industry Type | Forensics |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?