missing translation for 'onlineSavingsMsg'
Learn More

rac Indapamide-N-sulfonamido-Beta-D-glucuronide (Mixture of Diastereomers), TRC

Product Code. 30395583
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395583 10 mg 10mg
30397073 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395583 Supplier Toronto Research Chemicals Supplier No. TRCI50011510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Indapamide-N-sulfonamido-beta-D-glucuronide
Analyte or Component Names rac Indapamide-N-sulfonamido-beta-D-glucuronide
Shipping Condition Room Temperature
Molecular Formula C22 H24 Cl N3 O9 S
Synonym rac Indapamide-N-sulfonamido-β-D-glucuronide (Mixture of Diastereomers)
InChI Formula InChI=1S/C22H24ClN3O9S/c1-10-8-11-4-2-3-5-14(11)26(10)24-20(30)12-6-7-13(23)15(9-12)36(33,34)25-21-18(29)16(27)17(28)19(35-21)22(31)32/h2-7,9-10,16-19,21,25,27-29H,8H2,1H3,(H,24,30)(H,31,32)/t10?,16-,17-,18+,19-,21+/m0/s1
Solution Type Neat
SMILES CC1Cc2ccccc2N1NC(=O)c3ccc(Cl)c(c3)S(=O)(=O)N[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6R)-6-[[2-chloro-5-[(2-methyl-2,3-dihydroindol-1-yl)carbamoyl]phenyl]sulfonylamino]-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Forensics
Molecular Weight (g/mol) 541.96
Quantity 10 mg
Formula Weight 541.0922
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.