missing translation for 'onlineSavingsMsg'
Learn More

rac Ibuprofen-d3, TRC

Product Code. 30404637
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404637 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404637 Supplier Toronto Research Chemicals Supplier No. TRCI1400021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibuprofen-D3
CAS 121662-14-4
Purity Grade Notes HPLC
Molecular Formula C13 2H3 H15 O2
Quantity 1 mg
Synonym Benzeneacetic acid, α-(methyl-d3)-4-(2-methylpropyl)-, (±)-, α-(Methyl-d3)-4-(2-methylpropyl)benzeneacetic acid, Racibuprofen-d3, Benzeneacetic acid, α-(methyl-d3)-4-(2-methylpropyl)-, Ibuprofen D3 (Alpha-methyl D3)
InChI Formula InChI=1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)/i3D3
SMILES [2H]C([2H])([2H])C(C(=O)O)c1ccc(CC(C)C)cc1
IUPAC Name 3,3,3-trideuterio-2-[4-(2-methylpropyl)phenyl]propanoic acid
Molecular Weight (g/mol) 209.3
Formula Weight 209.1495
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.