missing translation for 'onlineSavingsMsg'
Learn More

rac Homatropine Hydrobromide, TRC

Product Code. 30399815
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399815 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399815 Supplier Toronto Research Chemicals Supplier No. TRCH4592002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Homatropine Hydrobromide
CAS 51-56-9
Purity Grade Notes HPLC
Molecular Formula C16 H21 N O3 . Br H
Quantity 2.5 g
Synonym Homatropine hydrobromide, (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl (2RS)-2-hydroxy-2-phenylacetate hydrobromide, 1αH,5αH-Tropan-3α-ol, mandelate (ester), hydrobromide (8CI), Benzeneacetic acid, α-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide (9CI), Benzeneacetic acid, α-hydroxy-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide, endo-, (±)-Homatropine bromide, Homatropine bromide, Homatropine hydrobromide, Isopto Homatropine, Mandelic acid tropine ester hydrobromide, Benzeneacetic acid, α-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide (1:1)
InChI Formula InChI=1S/C16H21NO3.BrH/c1-17-12-7-8-13(17)10-14(9-12)20-16(19)15(18)11-5-3-2-4-6-11;/h2-6,12-15,18H,7-10H2,1H3;1H/t12-,13+,14+,15-;/m1./s1
SMILES Br.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](O)c3ccccc3
IUPAC Name [(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;hydrobromide
Molecular Weight (g/mol) 356.25
Formula Weight 355.0783
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.