missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac Homatropine Hydrobromide, TRC
Specifications
Specifications
| CAS | 51-56-9 |
| Molecular Formula | C16 H21 N O3 . Br H |
| Quantity | 50 mg |
| Synonym | Homatropine hydrobromide, (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl (2RS)-2-hydroxy-2-phenylacetate hydrobromide, 1αH,5αH-Tropan-3α-ol, mandelate (ester), hydrobromide (8CI), Benzeneacetic acid, α-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide (9CI), Benzeneacetic acid, α-hydroxy-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide, endo-, (±)-Homatropine bromide, Homatropine bromide, Homatropine hydrobromide, Isopto Homatropine, Mandelic acid tropine ester hydrobromide, Benzeneacetic acid, α-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide (1:1) |
| InChI Formula | InChI=1S/C16H21NO3.BrH/c1-17-12-7-8-13(17)10-14(9-12)20-16(19)15(18)11-5-3-2-4-6-11;/h2-6,12-15,18H,7-10H2,1H3;1H/t12-,13+,14+,15-;/m1./s1 |
| SMILES | Br.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](O)c3ccccc3 |
| IUPAC Name | [(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;hydrobromide |
| Molecular Weight (g/mol) | 356.25 |
| Formula Weight | 355.0783 |
| Percent Purity | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?