missing translation for 'onlineSavingsMsg'
Learn More

rac Guaifenesin, TRC

Product Code. 30136909
Change view
Click to view available options
Quantity:
10 mg
1 g
25 g
Unit Size:
10mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136909 25 g 25g
17974565 10 mg 10mg
17931775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136909 Supplier Toronto Research Chemicals Supplier No. TRCG81050025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-14-1
Molecular Formula C10 H14 O4
Molecular Weight (g/mol) 198.22
Synonym 1,2-Propanediol, 3-(o-methoxyphenoxy)- (6CI,8CI), 3-(2-Methoxyphenoxy)-1,2-propanediol, 1,2-Dihydroxy-3-(2-methoxyphenoxy)propane, 2-G, 3-(o-Methoxyphenoxy)-1,2-propanediol, Actifed C, Aeronesin, Amonidren, Aresol, Calmipan, Colrex Expectorant, Creson, Dilyn, Equicol, Giafen, Glycerin guaiacolate, Glycerol guaiacolate, Glycerol α-(2-methoxyphenyl) ether, Glycerol α-(o-methoxyphenyl)ether, Glycerol α-guaiacyl ether, Glyceryl guaiacol ether, Glyceryl guaiacolate, Glyceryl guaiacolate ether, Glyceryl guaiacyl ether, Glycerylguaiacol, Glycodex, Glycotuss, Guaiacol glycerin ether, Guaiacol glycerol ether, Guaiacol glyceryl ether, Guaiacuran, Guaiacurane, Guaiacyl glyceryl ether, Guaiamar, Guaianesin, Guaifenesin, Guaifenesine, Guaiphenesin, Guaiphenesine, Guajacuran, Guanar, Guayanesin, Hustosil, Hytuss, Methphenoxydiol, Miocurin, Mucinex, Muskurelax, My 301, Myocain, Myocaine, Myoscain, NSC 62112, Neuroton, Neurotone, Oresol, Oreson, Organidin NR, Relaxil G, Reorganin, Respenyl, Resyl, Ritussin, Robitussin, SL 90, Sirotol, Tenntus, Tenntuss, Trecid, Tulyn, XL 90, o-Methoxyphenyl glyceryl ether, racemic-Guaifenesin, α-Glyceryl guaiacol ether, α-Glyceryl guaiacolate ether
IUPAC Name 3-(2-methoxyphenoxy)propane-1,2-diol
SMILES COc1ccccc1OCC(O)CO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-7-8(12)6-11/h2-5,8,11-12H,6-7H2,1H3
Formula Weight 198.0892
Chemical Name or Material Guaifenesin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.