missing translation for 'onlineSavingsMsg'
Learn More

rac Guaifenesin-d5 Cyclic Carbonate, TRC

Product Code. 30400620
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400620 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400620 Supplier Toronto Research Chemicals Supplier No. TRCG81051710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Guaifenesin-d5 Cyclic Carbonate
CAS 1329809-21-3
Molecular Formula C11H7D5O5
Quantity 10 mg
Synonym Carbonic Acid Cyclic [(o-Methoxyphenoxy)methyl]ethylene Ester-d5, 4-[(2-Methoxyphenoxy)methyl]-1,3-dioxolan-2-one-d5 3-(o-Methoxyphenoxy)-1,2-propanediol-d5 Cyclic Carbonate
InChI Formula InChI=1S/C11H12O5/c1-13-9-4-2-3-5-10(9)14-6-8-7-15-11(12)16-8/h2-5,8H,6-7H2,1H3/i6D2,7D2,8D
SMILES [2H]C([2H])(Oc1ccccc1OC)C2([2H])OC(=O)OC2([2H])[2H]
IUPAC Name 4,4,5-trideuterio-5-[dideuterio-(2-methoxyphenoxy)methyl]-1,3-dioxolan-2-one
Molecular Weight (g/mol) 229.24
Formula Weight 229.0999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.