missing translation for 'onlineSavingsMsg'
Learn More

(Rac)-GSK547, MedChemExpress

Product Code. 16497587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497587 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497587 Supplier Medchem Express Supplier No. HY114492A
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (Rac)-GSK547 is the racemate of GSK547. GSK547 is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1), inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer.

(Rac)-GSK547 is the racemate of GSK547. GSK547 is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1), inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer.

Specifications

Chemical Name or Material (Rac)-GSK547
Color White
Purity Grade Notes Research
Molecular Formula C20H18F2N6O
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (315.35 mM)
SMILES N#CC1=NC=NC(N2CCC(C(N3N=CCC3C4=CC(F)=CC(F)=C4)=O)CC2)=C1
Molecular Weight (g/mol) 396.39
Formula Weight 396.39
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.