missing translation for 'onlineSavingsMsg'
Learn More

rac Frovatriptan-d3 Succinate Monohydrate (2:1:1), TRC

Product Code. 30397035
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397035 10 mg 10mg
30403340 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30397035 Supplier Toronto Research Chemicals Supplier No. TRCF76850210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Frovatriptan-d3 Succinate Monohydrate (2:1:1)
Molecular Formula C14 D3 H14 N3 O . C4 H6 O4 . H2 O
Quantity 10 mg
Synonym rac-2,3,4,9-Tetrahydro-3-(methyl-d3-amino)-1H-carbazole-6-carboxamide Butanedioic Acid Monohydrate, Frova-d3, Frovelan-d3, Migard-d3, Miguard-d3, rac Difrovatriptan-d3 Succinate Monohydrate
InChI Formula InChI=1S/C14H17N3O.C4H6O4.H2O/c1-16-9-3-5-13-11(7-9)10-6-8(14(15)18)2-4-12(10)17-13;5-3(6)1-2-4(7)8;/h2,4,6,9,16-17H,3,5,7H2,1H3,(H2,15,18);1-2H2,(H,5,6)(H,7,8);1H2/i1D3;;
SMILES O.[2H]C([2H])([2H])NC1CCc2[nH]c3ccc(cc3c2C1)C(=O)N.OC(=O)CCC(=O)O
IUPAC Name butanedioic acid;6-(trideuteriomethylamino)-6,7,8,9-tetrahydro-5H-carbazole-3-carboxamide;hydrate
Molecular Weight (g/mol) 382.426
Formula Weight 382.193
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.