missing translation for 'onlineSavingsMsg'
Learn More

rac-Frovatriptan Acid, TRC

Product Code. 30402383
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402383 100 mg 100mg
30399879 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402383 Supplier Toronto Research Chemicals Supplier No. TRCF768510100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Frovatriptan Acid
Analyte or Component Names rac-Frovatriptan Acid
Shipping Condition Room Temperature
Molecular Formula C14 H16 N2 O2
Synonym 3-(Methylamino)-2,3,4,9-tetrahydro-1H-carbazole-6-carboxylic Acid
InChI Formula InChI=1S/C14H16N2O2/c1-15-9-3-5-13-11(7-9)10-6-8(14(17)18)2-4-12(10)16-13/h2,4,6,9,15-16H,3,5,7H2,1H3,(H,17,18)
Solution Type Neat
SMILES CNC1CCc2[nH]c3ccc(cc3c2C1)C(=O)O
IUPAC Name 6-(methylamino)-6,7,8,9-tetrahydro-5H-carbazole-3-carboxylic acid
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 244.29
Quantity 100 mg
Formula Weight 244.1212
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.