missing translation for 'onlineSavingsMsg'
Learn More

rac-Febrifugine Dihydrochloride, TRC

Product Code. 30394120
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394120 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394120 Supplier Toronto Research Chemicals Supplier No. TRCF2285005MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32434-42-7
Molecular Formula C16 H19 N3 O3 . 2 Cl H
Quantity 5 mg
Synonym 4(3H)-Quinazolinone, 3-[3-[(2R,3S)-3-hydroxy-2-piperidinyl]-2-oxopropyl]-, hydrochloride (1:2), 4(3H)-Quinazolinone, 3-[3-[(2R,3S)-3-hydroxy-2-piperidinyl]-2-oxopropyl]-, dihydrochloride (9CI), Febrifugine, dihydrochloride (8CI), (+)-Febrifugine dihydrochloride, 3-[3-[(2R,3S)-3-Hydroxy-2-piperidinyl]-2-oxopropyl]-(3H)-quinazolinone dihydrochloride
InChI Formula InChI=1S/C16H19N3O3.2ClH/c20-11(8-14-15(21)6-3-7-17-14)9-19-10-18-13-5-2-1-4-12(13)16(19)22;;/h1-2,4-5,10,14-15,17,21H,3,6-9H2;2*1H/t14-,15+;;/m1../s1
SMILES Cl.Cl.O[C@H]1CCCN[C@@H]1CC(=O)CN2C=Nc3ccccc3C2=O
IUPAC Name 3-[3-[(2R,3S)-3-hydroxypiperidin-2-yl]-2-oxopropyl]quinazolin-4-one;dihydrochloride
Molecular Weight (g/mol) 374.26
Formula Weight 373.096
Chemical Name or Material rac-Febrifugine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.