missing translation for 'onlineSavingsMsg'
Learn More

rac erythro-Dihydro Bupropion, TRC

Product Code. 30385176
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385176 5 mg 5mg
30387941 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385176 Supplier Toronto Research Chemicals Supplier No. TRCD4486505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99102-04-2
Molecular Formula C13 H20 Cl N O
Molecular Weight (g/mol) 241.76
Synonym Benzenemethanol, 3-chloro-α-[(1R)-1-[(1,1-dimethylethyl)amino]ethyl]-, (αS)-rel- (9CI, ACI), Benzenemethanol, 3-chloro-α-[1-[(1,1-dimethylethyl)amino]ethyl]-, (R*,S*)- (ZCI), rel-(αS)-3-Chloro-α-[(1R)-1-[(1,1-dimethylethyl)amino]ethyl]benzenemethanol (ACI), Erythrohydrobupropion, Erythrohydroxybupropion, rel-(1R,2S)-2-(tert-Butylamino)-1-(3-chlorophenyl)-1-propanol, rac erythro-Dihydro Bupropion, (±)-erythro-Dihydrobupropion
IUPAC Name (1R,2S)-2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-ol
SMILES C[C@H](NC(C)(C)C)[C@H](O)c1cccc(Cl)c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H20ClNO/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10/h5-9,12,15-16H,1-4H3/t9-,12-/m0/s1
Formula Weight 241.1233
Percent Purity >95%
Chemical Name or Material rac erythro-Dihydrobupropion
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.