missing translation for 'onlineSavingsMsg'
Learn More

rac erythro-9-(2-Hydroxy-3-nonyl)adenine Hydrochloride Salt, TRC

Product Code. 30397867
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397867 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397867 Supplier Toronto Research Chemicals Supplier No. TRCH8255501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac erythro-9-(2-Hydroxy-3-nonyl)adenine, Hydrochloride
CAS 58337-38-5
Molecular Formula C14 H23 N5 O . Cl H
Quantity 1 mg
Synonym 9H-Purine-9-ethanol, 6-amino-β-hexyl-α-methyl-, hydrochloride (1:1), (αR,βS)-rel- (ACI), 9H-Purine-9-ethanol, 6-amino-β-hexyl-α-methyl-, monohydrochloride, (R*,S*)- (9CI), erythro-9-(2-Hydroxy-3-nonyl)adenine hydrochloride, NSC 263164, erythro-9-(2-Hydroxy-3-nonyl)adenine monohydrochloride, (αR,βS)-rel-6-Amino-β-hexyl-α-methyl-9H-purine-9-ethanol monohydrochloride, rac erythro-9-(2-Hydroxy-3-nonyl)adenine hydrochloride salt
InChI Formula InChI=1S/C14H23N5O.ClH/c1-3-4-5-6-7-11(10(2)20)19-9-18-12-13(15)16-8-17-14(12)19;/h8-11,20H,3-7H2,1-2H3,(H2,15,16,17);1H/t10-,11+;/m0./s1
SMILES Cl.CCCCCC[C@H]([C@H](C)O)n1cnc2c(N)ncnc12
IUPAC Name (2S,3R)-3-(6-aminopurin-9-yl)nonan-2-ol;hydrochloride
Molecular Weight (g/mol) 313.83
Formula Weight 313.1669
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.