missing translation for 'onlineSavingsMsg'
Learn More

rac Enterolactone-d6, TRC

Product Code. 30377616
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377616 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30377616 Supplier Toronto Research Chemicals Supplier No. TRCE5589535MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Enterolactone-d6
CAS 104411-11-2
Molecular Formula C18H12D6O4
Quantity 5 mg
Synonym Dihydro-3,4-bis[(3-hydroxyphenyl)methyl]-2(3H)-furanone-d6, trans-Dihydro-3,4-bis[(3-hydroxyphenyl)methyl]-2(3H)-Furanone-d6, HPMF-d6, trans-2,3-Bis(3-hydroxybenzyl)-γ-butyrolactone-d6
InChI Formula InChI=1S/C18H18O4/c19-15-5-1-3-12(8-15)7-14-11-22-18(21)17(14)10-13-4-2-6-16(20)9-13/h1-6,8-9,14,17,19-20H,7,10-11H2/t14-,17+/m1/s1/i3D,4D,5D,6D,8D,9D
SMILES OC1=C([2H])C(C[C@H](C(OC2)=O)[C@@H]2CC3=C([2H])C=C([2H])C(O)=C3[2H])=C([2H])C=C1[2H]
Molecular Weight (g/mol) 304.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.