missing translation for 'onlineSavingsMsg'
Learn More

rac-Enprostil, TRC

Product Code. 30381912
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381912 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381912 Supplier Toronto Research Chemicals Supplier No. TRCE5577005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Enprostil
CAS 73121-56-9
Molecular Formula C23 H28 O6
Quantity 5 mg
Synonym Enprostil, 4,5-Heptadienoic acid, 7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3R)-3-hydroxy-4-phenoxy-1-buten-1-yl]-5-oxocyclopentyl]-, methyl ester, rel-, 4,5-Heptadienoic acid, 7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3R)-3-hydroxy-4-phenoxy-1-butenyl]-5-oxocyclopentyl]-, methyl ester, rel- (9CI), Camleed, Enprostil, Fundyl, Gardrin, Gardrine, RS 84-135, RS 84135, RS 84135-004, Syngard, rel-7-[(1R,2R,3R)-3-Hydroxy-2-[(1E,3R)-3-hydroxy-4-phenoxy-1-buten-1-yl]-5-oxocyclopentyl]-4,5-heptadienoic Acid Methyl Ester, rel-7-[(1R,2R,3R)-3-Hydroxy-2-[(1E,3R)-3-hydroxy-4-phenoxy-1-butenyl]-5-oxocyclopentyl]-4,5-heptadienoic Acid Methyl Ester
InChI Formula InChI=1S/C23H28O6/c1-28-23(27)12-8-3-2-7-11-19-20(22(26)15-21(19)25)14-13-17(24)16-29-18-9-5-4-6-10-18/h3-7,9-10,13-14,17,19-20,22,24,26H,8,11-12,15-16H2,1H3/b14-13+/t2?,17-,19-,20-,22-/m1/s1
SMILES COC(=O)CCC=C=CC[C@@H]1[C@@H](\C=C\[C@@H](O)COc2ccccc2)[C@H](O)CC1=O
IUPAC Name methyl 7-[(1R,2R,3R)-3-hydroxy-2-[(E,3R)-3-hydroxy-4-phenoxy-but-1-enyl]-5-oxo-cyclopentyl]hepta-4,5-dienoate
Molecular Weight (g/mol) 400.46
Formula Weight 400.1886
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.