missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac Duloxetine 3-Thiophene Isomer Maleic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | rac Duloxetine 3-Thiophene Isomer Maleic Acid Salt |
| Analyte or Component Names | rac Duloxetine 3-Thiophene Isomer Maleic Acid Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 N O S . C4 H4 O4 |
| InChI Formula | InChI=1S/C18H19NOS.C4H4O4/c1-19-11-9-17(15-10-12-21-13-15)20-18-8-4-6-14-5-2-3-7-16(14)18;5-3(6)1-2-4(7)8/h2-8,10,12-13,17,19H,9,11H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Solution Type | Neat |
| SMILES | CNCCC(Oc1cccc2ccccc12)c3ccsc3.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N-methyl-3-naphthalen-1-yloxy-3-thiophen-3-ylpropan-1-amine |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 413.487 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?