missing translation for 'onlineSavingsMsg'
Learn More

rac Desmethyl Citalopram Hydrobromide, TRC

Product Code. 30364993
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
100 mg
Unit Size:
100mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364993 25 mg 25mg
30354769 2.5 mg 2.5mg
30349150 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364993 Supplier Toronto Research Chemicals Supplier No. TRCD23087025MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188264-72-3
Molecular Formula C19 H19 F N2 O . Br H
Molecular Weight (g/mol) 391.28
Synonym 5-Isobenzofurancarbonitrile, 1-(4-fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-, hydrobromide (1:1), (1RS)-1-(4-Fluorophenyl)-1-[3-(methylamino)propyl]-1,3-dihydroisobenzofuran-5-carbonitrile Hydrobromide, Desmethylcitalopram Hydrobromide, rac Desmethyl Citalopram Hydrobromide
IUPAC Name 1-(4-fluorophenyl)-1-[3-(methylamino)propyl]-3H-2-benzofuran-5-carbonitrile;hydrobromide
SMILES Br.CNCCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H19FN2O.BrH/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;/h3-8,11,22H,2,9-10,13H2,1H3;1H
Formula Weight 390.0743
Percent Purity >95%
Chemical Name or Material rac Desmethyl Citalopram hydrobromide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.