missing translation for 'onlineSavingsMsg'
Learn More

rac-Crotonoyl Chloride, TRC

Product Code. 30367128
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367128 100 g 100g
30360886 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367128 Supplier Toronto Research Chemicals Supplier No. TRCC818795100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Crotonoyl Chloride
CAS 10487-71-5
Molecular Formula C4H5ClO
Quantity 100 g
Synonym Crotonic Acid Chloride, 2-Butenoyl Chloride, beta-Methylacryloyl Chloride
InChI Formula InChI=1S/C4H5ClO/c1-2-3-4(5)6/h2-3H,1H3
SMILES CC=CC(=O)Cl
IUPAC Name but-2-enoyl chloride
Molecular Weight (g/mol) 104.53
Formula Weight 104.0029
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.