missing translation for 'onlineSavingsMsg'
Learn More

rac-cis-Permethrinic Acid, TRC

Product Code. 30136614
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136614 1 g 1g
30132953 500 mg 500mg
17961875 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136614 Supplier Toronto Research Chemicals Supplier No. TRCP2877001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59042-49-8
Molecular Formula C8 H10 Cl2 O2
Molecular Weight (g/mol) 209.07
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, cis-, rel-(1R,3R)-3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic acid, (±)-cis-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid, (±)-cis-Cypermethric acid, (±)-cis-Permethrinic acid, cis-(Dichlorovinyl)dimethylcyclopropanecarboxylic acid, cis-(±)-Cypermethric acid, cis-2,2-Dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylic acid, cis-3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic acid, cis-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropane-1-carboxylic acid, cis-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid, cis-DL-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid, cis-Permethric acid, cis-Permethrinic acid, Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (1R,3R)-rel-
IUPAC Name (1S,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(C)[C@H](C=C(Cl)Cl)[C@@H]1C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H10Cl2O2/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3,(H,11,12)/t4-,6-/m1/s1
Formula Weight 208.0058
Chemical Name or Material rac-cis-Permethrinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.