missing translation for 'onlineSavingsMsg'
Learn More

rac-cis-Ambroxol, TRC

Product Code. 30133883
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133883 100 mg 100mg
30012191 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133883 Supplier Toronto Research Chemicals Supplier No. TRCA575905100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107814-37-9
Molecular Formula C13 H18 Br2 N2 O
Molecular Weight (g/mol) 378.1
Synonym Cyclohexanol, 4-[[(2-amino-3,5-dibromophenyl)methyl]amino]-, cis-, cis-4-[[(2-Amino-3,5-dibromophenyl)methyl]amino]cyclohexanol, cis-Ambroxol, cis-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol
IUPAC Name 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol
SMILES Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@H](O)CC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H18Br2N2O/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10/h5-6,10-11,17-18H,1-4,7,16H2/t10-,11+
Formula Weight 375.9786
Percent Purity >95%
Chemical Name or Material cis-Ambroxol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.