missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac cis-9,10-Epoxystearic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | rac cis-9,10-Epoxystearic Acid |
| CAS | 24560-98-3 |
| Purity Grade Notes | GC |
| Molecular Formula | C18 H34 O3 |
| Quantity | 100 mg |
| Synonym | (2R,3S)-rel-3-Octyl-2-oxiraneoctanoic Acid, cis-9,10-Epoxyoctadecanoic Acid, (2R,3S)-rel-3-Octyloxiraneoctanoic Acid, cis-3-Octyloxiraneoctanoic Acid, cis-9,10-Epoxyoctadecanoate, cis-9,10-Epoxyoctadecanoic Acid, cis-9,10-Epoxystearic Acid, cis-DL-9,10-Epoxystearic Acid, cis-Epoxystearic Acid |
| InChI Formula | InChI=1S/C18H34O3/c1-2-3-4-5-7-10-13-16-17(21-16)14-11-8-6-9-12-15-18(19)20/h16-17H,2-15H2,1H3,(H,19,20)/t16-,17+/m0/s1 |
| SMILES | CCCCCCCC[C@@H]1O[C@@H]1CCCCCCCC(=O)O |
| IUPAC Name | 8-[(2R,3S)-3-octyloxiran-2-yl]octanoic acid |
| Molecular Weight (g/mol) | 298.46 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?