missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac cis-3-Hydroxy Glyburide (Methoxy-13CD3), TRC
Specifications
Specifications
| Chemical Name or Material | rac cis-3-Hydroxy Glyburide-d3,13C |
| CAS | 1217848-91-3 |
| Molecular Formula | 13C C22 D3 H25 Cl N3 O6 S |
| Quantity | 0.25 mg |
| Synonym | Rac Cis-3-Hydroxy Glyburide 13C,D3 |
| InChI Formula | InChI=1S/C23H28ClN3O6S/c1-33-21-10-7-16(24)13-20(21)22(29)25-12-11-15-5-8-19(9-6-15)34(31,32)27-23(30)26-17-3-2-4-18(28)14-17/h5-10,13,17-18,28H,2-4,11-12,14H2,1H3,(H,25,29)(H2,26,27,30)/t17-,18+/m0/s1/i1+1D3 |
| SMILES | [2H][13C]([2H])([2H])Oc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)N[C@H]3CCC[C@@H](O)C3 |
| IUPAC Name | 5-chloro-N-[2-[4-[[(1S,3R)-3-hydroxycyclohexyl]carbamoylsulfamoyl]phenyl]ethyl]-2-(trideuterio(113C)methoxy)benzamide |
| Molecular Weight (g/mol) | 514.014 |
| Formula Weight | 513.161 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?