missing translation for 'onlineSavingsMsg'
Learn More

rac Cetirizine N-Oxide >90% by HPLC(Mixture of Diastereomers), TRC

Product Code. 30136218
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136218 100 mg 100mg
30134971 25 mg 25mg
17970795 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136218 Supplier Toronto Research Chemicals Supplier No. TRCC281110100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076199-80-8
Molecular Formula C21 H25 Cl N2 O4
Molecular Weight (g/mol) 404.89
Synonym Cetirizine N-Oxide, 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]-1-oxido-1-piperazinyl]ethoxy]acetic acid, Cetirizine-N-oxide
IUPAC Name 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]-1-oxidopiperazin-1-ium-1-yl]ethoxy]acetic acid
SMILES OC(=O)COCC[N+]1([O-])CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H25ClN2O4/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)23-10-12-24(27,13-11-23)14-15-28-16-20(25)26/h1-9,21H,10-16H2,(H,25,26)
Formula Weight 404.1503
Chemical Name or Material Cetirizine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.