missing translation for 'onlineSavingsMsg'
Learn More

rac-Alpha-[2-(Dimethylamino)ethyl]-3-thiophenemethanol, TRC

Product Code. 30383957
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383957 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383957 Supplier Toronto Research Chemicals Supplier No. TRCD471825100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-alpha-[2-(Dimethylamino)ethyl]-3-thiophenemethanol
CAS 116817-84-6
Molecular Formula C9H15NOS
Quantity 100 mg
Synonym alpha-[2-(Dimethylamino)ethyl]-3-thiophenemethanol
InChI Formula InChI=1S/C9H15NOS/c1-10(2)5-3-9(11)8-4-6-12-7-8/h4,6-7,9,11H,3,5H2,1-2H3
SMILES CN(C)CCC(O)c1ccsc1
Molecular Weight (g/mol) 185.29
Formula Weight 185.0874
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.