missing translation for 'onlineSavingsMsg'
Learn More

rac all-trans 3-(Acetyloxy) Retinol Acetate 90%, TRC

Product Code. 30400938
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400938 1 mg 1mg
30400835 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400938 Supplier Toronto Research Chemicals Supplier No. TRCH9533051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material all-trans 3-Hydroxyretinol
CAS 76686-33-4
Molecular Formula C24H34O4
Quantity 1 mg
Synonym Rac All-Trans 3-(Acetyloxy) Retinol Acetate
InChI Formula InChI=1S/C24H34O4/c1-17(9-8-10-18(2)13-14-27-20(4)25)11-12-23-19(3)15-22(28-21(5)26)16-24(23,6)7/h8-13,22H,14-16H2,1-7H3/b10-8+,12-11+,17-9+,18-13+
SMILES CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CC(CC1(C)C)OC(=O)C
IUPAC Name [(2E,4E,6E,8E)-9-(4-acetyloxy-2,6,6-trimethylcyclohexen-1-yl)-3,7-dimethylnona-2,4,6,8-tetraenyl] acetate
Molecular Weight (g/mol) 386.52
Formula Weight 386.2457
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.