missing translation for 'onlineSavingsMsg'
Learn More

rac 7-Hydroxy Propranolol, TRC

Product Code. 30407173
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407173 1 mg
30402949 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407173 Supplier Toronto Research Chemicals Supplier No. TRCH9525551MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81907-81-5
Molecular Formula C16 H21 N O3
Molecular Weight (g/mol) 275.34
IUPAC Name 8-[2-hydroxy-3-(propan-2-ylamino)propoxy]naphthalen-2-ol
SMILES CC(C)NCC(O)COc1cccc2ccc(O)cc12

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H21NO3/c1-11(2)17-9-14(19)10-20-16-5-3-4-12-6-7-13(18)8-15(12)16/h3-8,11,14,17-19H,9-10H2,1-2H3
Formula Weight 275.1521
Chemical Name or Material rac 7-Hydroxy Propranolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.