missing translation for 'onlineSavingsMsg'
Learn More

rac-5-Methylcotinine, TRC

Product Code. 30482452
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482452 25 mg 25mg
30476080 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482452 Supplier Toronto Research Chemicals Supplier No. TRCM29452525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076198-50-9
Molecular Formula C11 H14 N2 O
Molecular Weight (g/mol) 190.24
IUPAC Name 1-methyl-5-(5-methylpyridin-3-yl)pyrrolidin-2-one
SMILES CN1C(CCC1=O)c2cncc(C)c2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H14N2O/c1-8-5-9(7-12-6-8)10-3-4-11(14)13(10)2/h5-7,10H,3-4H2,1-2H3
Formula Weight 190.1106
Chemical Name or Material (+/-)-5-Methylcotinine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.