missing translation for 'onlineSavingsMsg'
Learn More

rac-4’-Methyl Ketoprofen, TRC

Product Code. 30481182
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481182 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481182 Supplier Toronto Research Chemicals Supplier No. TRCM31508050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2-(3-(4-Methylbenzoyl)phenyl)propanoic Acid
CAS 107257-20-5
Purity Grade Notes HPLC
Molecular Formula C17 H16 O3
Quantity 50 mg
Synonym (2RS)-2-[3-(4-Methylbenzoyl)phenyl]propanoic Acid, Ketoprofen Imp. D (EP)
InChI Formula InChI=1S/C17H16O3/c1-11-6-8-13(9-7-11)16(18)15-5-3-4-14(10-15)12(2)17(19)20/h3-10,12H,1-2H3,(H,19,20)
SMILES CC(C(=O)O)c1cccc(c1)C(=O)c2ccc(C)cc2
IUPAC Name 2-[3-(4-methylbenzoyl)phenyl]propanoic acid
Molecular Weight (g/mol) 268.31
Formula Weight 268.1099
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.