missing translation for 'onlineSavingsMsg'
Learn More

rac-4-Keto Frovatriptan, TRC

Product Code. 30404108
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404108 25 mg 25mg
30399400 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30404108 Supplier Toronto Research Chemicals Supplier No. TRCK18892025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-4-Keto Frovatriptan
Analyte or Component Names rac-4-Keto Frovatriptan
Shipping Condition Room Temperature
Molecular Formula C14 H15 N3 O2
Synonym 3-(Methylamino)-4-oxo-2,3,4,9-tetrahydro-1H-carbazole-6-carboxamide
InChI Formula InChI=1S/C14H15N3O2/c1-16-11-5-4-10-12(13(11)18)8-6-7(14(15)19)2-3-9(8)17-10/h2-3,6,11,16-17H,4-5H2,1H3,(H2,15,19)
Solution Type Neat
SMILES CNC1CCc2[nH]c3ccc(cc3c2C1=O)C(=O)N
IUPAC Name 6-(methylamino)-5-oxo-6,7,8,9-tetrahydrocarbazole-3-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 257.288
Quantity 25 mg
Formula Weight 257.116
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.