missing translation for 'onlineSavingsMsg'
Learn More

rac 4-Hydroxy-4-(3-pyridyl)butanoic Acid Dicyclohexylamine Salt, TRC

Product Code. 30137173
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137173 50 mg 50mg
30008165 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137173 Supplier Toronto Research Chemicals Supplier No. TRCH95297050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216491-66-5
Molecular Formula C21H34N2O3
Molecular Weight (g/mol) 362.51
SMILES OC(CCC(=O)O)c1cccnc1.C2CCC(CC2)NC3CCCCC3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H23N.C9H11NO3/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;11-8(3-4-9(12)13)7-2-1-5-10-6-7/h11-13H,1-10H2;1-2,5-6,8,11H,3-4H2,(H,12,13)
Formula Weight 362.2569
Percent Purity >95%
Chemical Name or Material (+/-)-4-Hydroxy-4-(3-pyridyl)butanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.