missing translation for 'onlineSavingsMsg'
Learn More

rac 3-Hydroxybutyric Acid Sodium Salt, TRC

Product Code. 17998574
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998574 10 g 10g
17938494 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17998574 Supplier Toronto Research Chemicals Supplier No. TRCH83302010G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-83-4
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.09
Synonym 3-Hydroxybutanoic Acid Sodium Salt, 3-Hydroxybutyric Acid Monosodium Salt, DL-β-Hydroxybutyric Acid Sodium Salt, Sodium 3-Hydroxybutyrate, Sodium Dl-β-hydroxybutyrate, Sodium β-Hydroxybutyrate, β-Hydroxybutyric Acid Sodium Salt;
SMILES [Na+].CC(O)CC(=O)[O-]

Specifications

Quantity 10 g
Chemical Name or Material rac 3-Hydroxybutyric Acid Sodium Salt (BHB)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.