missing translation for 'onlineSavingsMsg'
Learn More

rac- (2E)-3-[(Ethoxycarbonyl)methylene]-1-trityl-4-piperidinol, TRC

Product Code. 30382812
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382812 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382812 Supplier Toronto Research Chemicals Supplier No. TRCE89052525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac- (2E)-3-[(Ethoxycarbonyl)methylene]-1-trityl-4-piperidinol
CAS 239466-40-1
Molecular Formula C28H29NO3
Quantity 25 mg
Synonym (2E)-[4-Hydroxy-1-(triphenylmethyl)-3-piperidinylidene]acetic Acid Ethyl Ester, (2E)-3-[(Ethoxycarbonyl)methylene]-1-(triphenylmethyl)piperidin-4-ol
InChI Formula InChI=1S/C28H29NO3/c1-2-32-27(31)20-22-21-29(19-18-26(22)30)28(23-12-6-3-7-13-23,24-14-8-4-9-15-24)25-16-10-5-11-17-25/h3-17,20,26,30H,2,18-19,21H2,1H3/b22-20+
SMILES CCOC(=O)\C=C\1/CN(CCC1O)C(c2ccccc2)(c3ccccc3)c4ccccc4
IUPAC Name ethyl (2E)-2-(4-hydroxy-1-tritylpiperidin-3-ylidene)acetate
Molecular Weight (g/mol) 427.53
Formula Weight 427.2147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.