missing translation for 'onlineSavingsMsg'
Learn More

rac 1-Palmitoyl-3-chloropropanediol, TRC

Product Code. 30427221
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427221 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427221 Supplier Toronto Research Chemicals Supplier No. TRCP15650025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Palmitoyl-3-chloropropanediol
CAS 30557-04-1
Purity Grade Notes HPLC
Molecular Formula C19 H37 Cl O3
Quantity 25 mg
Synonym Palmitic acid','3-chloro-2-hydroxypropyl ester (8CI), 3-Chloro-1,2-propanediol 1-palmitate, Rac-1-palmitoyl-3-chloropropane-1,2-diol
InChI Formula InChI=1S/C19H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h18,21H,2-17H2,1H3
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)CCl
IUPAC Name (3-chloro-2-hydroxypropyl) hexadecanoate
Molecular Weight (g/mol) 348.95
Formula Weight 348.2431
Percent Purity >95
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.