missing translation for 'onlineSavingsMsg'
Learn More

rac 1-Palmitoyl-2-chloropropanediol, TRC

Product Code. 30138605
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138605 10 mg 10mg
30135971 5 mg 5mg
30004573 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138605 Supplier Toronto Research Chemicals Supplier No. TRCP15649010MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63326-63-6
Molecular Formula C19H37ClO3
Molecular Weight (g/mol) 348.95
Synonym Hexadecanoic Acid 2-Chloro-3-hydroxypropyl Ester;
IUPAC Name [(2S)-2-chloro-3-hydroxypropyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC[C@H](CO)Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(20)16-21/h18,21H,2-17H2,1H3/t18-/m0/s1
Formula Weight 348.2431
Chemical Name or Material rac 1-Palmitoyl-2-chloropropanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.