missing translation for 'onlineSavingsMsg'
Learn More

rac 1-Oleoyl-3-chloropropanediol, TRC

Product Code. 30137418
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137418 50 mg 50mg
30133743 25 mg 25mg
17988974 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137418 Supplier Toronto Research Chemicals Supplier No. TRCO52680050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10311-82-7
Molecular Formula C21 H39 Cl O3
Molecular Weight (g/mol) 374.99
Synonym 9-Octadecenoic acid (Z)-, 3-chloro-2-hydroxypropyl ester, Oleic acid, 3-chloro-2-hydroxypropyl ester (7CI,8CI), 1,2-Propanediol, 3-chloro-, 1-oleate (8CI), 3-Chloro-1,2-propanediol 1-oleate, 3-Chloro-1,2-propanediol, 1-monooleate
IUPAC Name (3-chloro-2-hydroxypropyl) (Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CCl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H39ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h9-10,20,23H,2-8,11-19H2,1H3/b10-9-
Formula Weight 374.2588
Percent Purity >95%
Chemical Name or Material 1-Oleoyl-3-chloropropanediol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.