missing translation for 'onlineSavingsMsg'
Learn More

rac-1,2-Dilinolenoyl-3-chloropropanediol, TRC

Product Code. 30387599
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387599 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387599 Supplier Toronto Research Chemicals Supplier No. TRCD46056525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-1,2-Dilinolenoyl-3-chloropropanediol
CAS 74875-97-1
Molecular Formula C39 H63 Cl O4
Quantity 25 mg
Synonym 9,12,15-Octadecatrienoic acid, 1-(chloromethyl)-1,2-ethanediyl ester, (all-Z)- (9CI), 3-Chloro-1,2-propanediol dilinolenate, Rac-1,2-bis(linolenoyl)-3-chloropropane-1,2-diol
InChI Formula InChI=1S/C39H63ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h5-8,11-14,17-20,37H,3-4,9-10,15-16,21-36H2,1-2H3/b7-5-,8-6-,13-11-,14-12-,19-17-,20-18-
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OCC(CCl)OC(=O)CCCCCCC\C=C/C\C=C/C\C=C/CC
IUPAC Name [3-chloro-2-[(9Z,12Z,15Z)-octadeca-9,12,15-trienoyl]oxypropyl] (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Molecular Weight (g/mol) 631.37
Formula Weight 630.4415
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.