missing translation for 'onlineSavingsMsg'
Learn More

rac-1, 2-Didecanoyl-3-chloropropanediol, TRC

Product Code. 30388200
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388200 250 mg 250mg
30371888 10 mg 10mg
30381520 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388200 Supplier Toronto Research Chemicals Supplier No. TRCD439800250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-1, 2-Didecanoyl-3-chloropropanediol
Analyte or Component Names rac-1, 2-Didecanoyl-3-chloropropanediol
Shipping Condition Room Temperature
Molecular Formula C23H43ClO4
Synonym rac-1, 2-Dicaprinoyl-3-chloropropanediol
InChI Formula InChI=1S/C23H43ClO4/c1-3-5-7-9-11-13-15-17-22(25)27-20-21(19-24)28-23(26)18-16-14-12-10-8-6-4-2/h21H,3-20H2,1-2H3
Solution Type Neat
SMILES CCCCCCCCCC(OCC(CCl)OC(CCCCCCCCC)=O)=O
Industry Type Forensics
Molecular Weight (g/mol) 419.04
Quantity 250 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.