missing translation for 'onlineSavingsMsg'
Learn More

rac 1,2-Diaminopropane, TRC

Product Code. 30040933
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040933 1 g 1g
30041270 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30040933 Supplier Toronto Research Chemicals Supplier No. TRCD4166551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-90-0
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.12
Synonym (RS)-1,2-Propanediamine, (±)-1,2-Diaminopropane, 1,2-Diaminopropane, 1,2-Propylenediamine, 1-Methyl-1,2-diaminoethane, 2,3-Diaminopropane, DL-1,2-Propanediamine, N,N'-Dimethyl-1,2-diaminopropane, NSC 175731, Propylenediamine, dl-1,2-Propylenediamine
SMILES NCC(N)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H10N2/c1-3(5)2-4/h3H,2,4-5H2,1H3
Formula Weight 74.0844
Chemical Name or Material rac 1,2-Diaminopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.