missing translation for 'onlineSavingsMsg'
Learn More

rac-1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol, TRC

Product Code. 30133129
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133129 10 g 10g
18051030 250 mg 250mg
18072920 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133129 Supplier Toronto Research Chemicals Supplier No. TRCD43579510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24155-42-8
Molecular Formula C11 H10 Cl2 N2 O
Molecular Weight (g/mol) 257.12
Synonym Imidazole-1-ethanol, α-(2,4-dichlorophenyl)- (8CI), α-(2,4-Dichlorophenyl)-1H-imidazole-1-ethanol, (±)-α-(2,4-Dichlorophenyl)-1H-imidazole-1-ethanol, 1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol, 1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethan-1-ol, 1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol, FI 7001, R 14821, T 824, UK 11147, (1RS)-1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol: Econazole Imp. A (EP)
IUPAC Name 1-(2,4-dichlorophenyl)-2-imidazol-1-ylethanol
SMILES OC(Cn1ccnc1)c2ccc(Cl)cc2Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H10Cl2N2O/c12-8-1-2-9(10(13)5-8)11(16)6-15-4-3-14-7-15/h1-5,7,11,16H,6H2
Formula Weight 256.017
Percent Purity >95%
Chemical Name or Material 1-(2,4-Dichlorophenyl)-2-imidazol-1-yl ethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.